Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydrazino-4-(2-thienyl)thiazole, 97%, Thermo Scientific™

Catalog No. AAH5829303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5829303 1 g
AAH5829306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5829303 Supplier Thermo Scientific Chemicals Supplier No. H5829303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21036-68-0
Molecular Formula C7H7N3S2
Molecular Weight (g/mol) 197.27
MDL Number MFCD06739917
InChI Key QYFFQQHJTLJVGU-UHFFFAOYSA-N
Synonym 4-thiophen-2-yl-thiazol-2-yl-hydrazine, 2-hydrazinyl-4-thiophen-2-yl-1,3-thiazole, 2-hydrazino-4-2-thienyl thiazole, 4-thiophen-2-ylthiazol-2-yl hydrazine, 2-hydrazinyl-4-thiophen-2-yl thiazole, 4-thiophen-2-yl-1,3-thiazol-2-yl hydrazine
PubChem CID 7174026
SMILES NNC1=NC(=CS1)C1=CC=CS1

Specifications

Quantity 1 g
Beilstein 1570002
IUPAC Name 2-hydrazinyl-4-(thiophen-2-yl)-1,3-thiazole
Formula Weight 197.27
Percent Purity 97%
Chemical Name or Material 2-Hydrazino-4-(2-thienyl)thiazole
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.