Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydrazinopyrazine, 98%

Catalog No. AAH64086MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64086MD 250 mg
AAH6408603 1 g
AAH6408606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64086MD Supplier Thermo Scientific Chemicals Supplier No. H64086MD

CAS: 54608-52-5 | C4H6N4 | 110.12 g/mol

Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triazolo[4,3-a]pyrazine 7-oxide, 3-β-D-ribofuranosyl-1,2,4-triazolo[4,3-a]pyrazine, monohydrazones of pyridine, pyrazine, phenylglyoxal were synthesised from 2-Hydrazinopyrazine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54608-52-5
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.12
MDL Number MFCD04114555
InChI Key IVRLZJDPKUSDCF-UHFFFAOYSA-N
Synonym 2-hydrazinylpyrazine, 2-hydrazinopyrazine, pyrazin-2-yl-hydrazine, 1-pyrazin-2-yl hydrazine, hydrazinopyrazine, 2-pyrazin-2-yl hydrazine, 2 1h-pyrazinone, hydrazone, hydrazino pyrazine, pyrazinyl hydrazine, 3-hydrazinopyrazine
PubChem CID 1487823
IUPAC Name pyrazin-2-ylhydrazine
SMILES C1=CN=C(C=N1)NN

Specifications

Melting Point 109°C to 113°C
Color Yellow
Quantity 250 mg
Beilstein 606630
Solubility Information Slightly soluble in water.
Formula Weight 110.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Hydrazinopyrazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.