Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-2-azaadamantane 98.0+%, TCI America™
SDP

Catalog No. H14041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14041G 1 g
H1404200MG 200 mg
H14045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H14041G Supplier TCI America Supplier No. H14041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1155843-79-0
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.225
InChI Key SFXINVPZIZMFHM-UHFFFAOYSA-N
Synonym AZADOL(regR)
PubChem CID 51037224
SMILES C1C2CC3CC1CC(C2)N3O

Specifications

Quantity 1 g
Formula Weight 153.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-2-azaadamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.