Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-2-methylpropiophenone 96.0+%, TCI America™
SDP

Catalog No. H0991100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0991100G 100 g
H099125G 25 g
H0991500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0991100G Supplier TCI America Supplier No. H0991100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7473-98-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00040710
InChI Key XMLYCEVDHLAQEL-UHFFFAOYSA-N
Synonym 2-Benzoyl-2-propanol
PubChem CID 81984
IUPAC Name 2-hydroxy-2-methyl-1-phenylpropan-1-one
SMILES CC(C)(C(=O)C1=CC=CC=C1)O

Specifications

Color Yellow
Boiling Point 110°C
Quantity 100 g
Formula Weight 164.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-2-methylpropiophenone
EINECSNumber (3)-3581
RTECSNumber US2977245
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.