Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-3-butynoic Acid 97.0+%, TCI America™
SDP

Catalog No. H0905100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0905100MG 100 mg
H09051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0905100MG Supplier TCI America Supplier No. H0905100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38628-65-8
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.073
MDL Number MFCD00671546
InChI Key LWNHDEQKHFRYMD-UHFFFAOYSA-N
PubChem CID 170267
IUPAC Name 2-hydroxybut-3-ynoic acid
SMILES C#CC(C(=O)O)O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 100.07
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-3-butynoic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.