Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-3-methylpyrazine 98.0+%, TCI America™
SDP

Catalog No. H15191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15191G 1 g
H15195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15191G Supplier TCI America Supplier No. H15191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19838-07-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD01646120
InChI Key LDQRWMQHTORUIY-UHFFFAOYSA-N
PubChem CID 88276
IUPAC Name 3-methyl-1H-pyrazin-2-one
SMILES CC1=NC=CNC1=O

Specifications

Melting Point 150°C
Color Yellow
Quantity 1 g
Synonym 2-hydroxy-3-methylpyrazine, 3-methylpyrazin-2-ol, 3-methylpyrazin-2 1h-one, 3-methyl-2-pyrazinol, 3-methyl-2 1h-pyrazinone, unii-s951qmj75e, 2 1h-pyrazinone, 3-methyl, 3-methyl-2-hydroxypyrazine, 2 1h-pyrazinone,3-methyl
Formula Weight 110.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-3-methylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.