Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4,6-dimethoxybenzoic acid, 97%, Thermo Scientific™

Catalog No. AAL0138806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0138806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0138806 Supplier Thermo Scientific Chemicals Supplier No. L0138806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Hydroxy-4,6-dimethoxybenzoic acid is a reagent used in the synthesis of rac Zearalenone-d6. rac Zearalenone-d6 s an estrogenic mycotoxin produced by Fusarium fungi commonly found in grains. One of a group of compounds known as resorcylic acid lactones.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3187-19-7
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00013977
InChI Key LEIJLTBRQHFMDL-UHFFFAOYSA-N
Synonym 4,6-dimethoxysalicylic acid, 4,6-dimethoxy-2-hydroxybenzoic acid, benzoic acid, 2-hydroxy-4,6-dimethoxy, acmc-1covk, 2-hydroxy-4,6-dimethoxybenzoicacid, 2,4-dimethoxy-6-hydroxybenzoic acid, 2-hydroxy-4,6-dimethoxy benzoic acid, 2-hydroxy-4,6-dimethoxy-benzoic acid, benzoic acid,2-hydroxy-4,6-dimethoxy
PubChem CID 76656
IUPAC Name 2-hydroxy-4,6-dimethoxybenzoic acid
SMILES COC1=CC(=C(C(=C1)OC)C(=O)O)O

Specifications

Melting Point 160°C to 162°C
Quantity 5 g
Beilstein 2213838
Solubility Information Insoluble in water.
Formula Weight 198.17
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-4,6-dimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-683-3
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.