Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxy-4-methoxybenzophenone

Catalog No. H36206100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H36206100G 100 g
H362065G 5 g
H36206500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H36206100G Supplier Sigma Aldrich Supplier No. H36206100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxy-4-methoxybenzophenone

Specifications

CAS 131-57-7
Melting Point 62°C to 64°C (lit.)
Boiling Point 150°C to 160°C (5 mmHg)
Molecular Formula C14H12O3
Linear Formula HOC6H3(OCH3)COC6H5
MDL Number MFCD00008387
Quantity 100 g
Synonym Oxybenzone
Molecular Weight (g/mol) 228.24
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.