Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-5'-methyl-3'-nitroacetophenone 98.0+%, TCI America™
SDP

Catalog No. H147925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H147925G 25 g
H14795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H147925G Supplier TCI America Supplier No. H147925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66108-30-3
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00192216
InChI Key XSHQMMIEZHWNAK-UHFFFAOYSA-N
Synonym 2'-hydroxy-5'-methyl-3'-nitroacetophenone, 1-2-hydroxy-5-methyl-3-nitrophenyl ethanone, 2-hydroxy-5-methyl-3-nitroacetophenone, 2-acetyl-4-methyl-6-nitrophenol, ethanone,1-2-hydroxy-5-methyl-3-nitrophenyl, 1-acetyl-2-hydroxy-5-methyl-3-nitrobenzene, 2'-hydroxy-3'-nitro-5'-methylacetophenone, 1-2-hydroxy-5-methyl-3-nitro-phenyl-ethanone, 1-5-methyl-3-nitro-2-oxidanyl-phenyl ethanone, 1-2-hydroxy-5-methyl-3-nitrophenyl-1-ethanone
PubChem CID 736162
IUPAC Name 1-(2-hydroxy-5-methyl-3-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC(C)=CC(=C1O)[N+]([O-])=O

Specifications

Melting Point 134°C
Color Yellow
Quantity 25 g
Formula Weight 195.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-5′-methyl-3′-nitroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.