Learn More
Cayman Chemical a-ethyl 2C-DhydrochlorIde 1mg
SDP
Supplier:  Cayman Chemical 265961 MG
Encompass_Preferred
A nucleoside analog inhibits the replication of murine S180 lung carcinoma cells and Ehrlich ascites mammary carcinoma cells (ED50s 8.5 and 4 M respectively) and multiple human leukemia cell lines (IC50s 1.7-5.8 M) acts synergistically with 5-FU against HT-29 HCT116 PANC-1 and EKVX cancer cells inhibits HSV-1 pyrimidine 2-deoxyribonucleoside kinase (Ki 3.5 M) and reduces HSV-1 viral titer to 0.05 of the control at a concentration of 200 M a DNA adduct formed in response to oxidative stress
Catalog No. 50-293-3903
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.