Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. H0192100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0192100G 100 g
H019225G 25 g
H0192500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0192100G Supplier TCI America Supplier No. H0192100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 118-93-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002219
InChI Key JECYUBVRTQDVAT-UHFFFAOYSA-N
Synonym 2'-hydroxyacetophenone, 2-acetylphenol, 1-2-hydroxyphenyl ethanone, o-hydroxyacetophenone, o-acetylphenol, o-hydroxyphenyl methyl ketone, ethanone, 1-2-hydroxyphenyl, 1-2-hydroxyphenyl ethan-1-one, acetophenone, 2'-hydroxy, 2-hydroxyphenyl methyl ketone
PubChem CID 8375
IUPAC Name 1-(2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1O

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 218°C
Quantity 100 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 2′-Hydroxyacetophenone
RTECSNumber AM8575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.