Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. H034725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H034725G 25 g
H03475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H034725G Supplier TCI America Supplier No. H034725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 582-24-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00041829
InChI Key ZWVHTXAYIKBMEE-UHFFFAOYSA-N
Synonym 2-hydroxyacetophenone, benzoylcarbinol, glycolophenone, phenacyl alcohol, alpha-hydroxyacetophenone, 2-hydroxy-1-phenylethan-1-one, ethanone, 2-hydroxy-1-phenyl, methanol, benzoyl, omega-hydroxyacetophenone, acetophenone, 2-hydroxy
PubChem CID 68490
ChEBI CHEBI:28341
IUPAC Name 2-hydroxy-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CO

Specifications

Melting Point 88°C
Color White-Yellow
Boiling Point 126°C
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyacetophenone
EINECSNumber (3)-4208
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.