Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl Disulfide 95%, Thermo Scientific™

Catalog No. AC176240250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176240250 25 g
AC176241000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC176240250 Supplier Thermo Scientific Chemicals Supplier No. 176240250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1892-29-1
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00002906
InChI Key KYNFOMQIXZUKRK-UHFFFAOYSA-N
Synonym 2-hydroxyethyl disulfide, 2,2'-dithiodiethanol, bis 2-hydroxyethyl disulfide, ethanol, 2,2'-dithiobis, hydroxyethyl disulfide, diethanol disulfide, 2,2'-dithiobisethanol, 2-hydroxyethanedisulfide, usaf th-9, 2,2'-disulfanediyldiethanol
PubChem CID 15906
ChEBI CHEBI:43136
IUPAC Name 2-[(2-hydroxyethyl)disulfanyl]ethan-1-ol
SMILES OCCSSCCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 1.26
Infrared Spectrum Authentic
Refractive Index 1.5650 to 1.5710 (20°C, 589nm)
Linear Formula (HOCH2CH2)2S2
Quantity 25 g
Beilstein 01, 471
Formula Weight 154.25
Percent Purity ≥94%
Physical Form Viscous Liquid After Melting
Chemical Name or Material 2-Hydroxyethyl disulfide, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 217-576-6
RTECSNumber KK7525000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.