Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyquinoxaline 98.0+%, TCI America™
SDP

Catalog No. H059125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H059125G 25 g
H0591100G 100 g
H0591500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H059125G Supplier TCI America Supplier No. H059125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1196-57-2
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006722
InChI Key FFRYUAVNPBUEIC-UHFFFAOYSA-N
Synonym 2-hydroxyquinoxaline, quinoxalin-2-ol, 2-quinoxalinol, 2 1h-quinoxalinone, quinoxalin-2 1h-one, quinoxalin-2-one, 2-quinoxalinone, 3-quinoxalinone, 2-quinoxalone, 1,2-dihydroquinoxalin-2-one
PubChem CID 14526
ChEBI CHEBI:38890
IUPAC Name 1,2-dihydroquinoxalin-2-one
SMILES O=C1NC2=CC=CC=C2N=C1

Specifications

Melting Point 275°C
Quantity 25 g
Formula Weight 146.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyquinoxaline
EINECSNumber (5)-6201
RTECSNumber VD3630000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.