Learn More
Eppendorf 3P Freezer Rack: CryoCube FC660 series, 2 in/53 mm, side access, stainless steel
Supplier:  Eppendorf 3P 6001050210
Encompass_Preferred
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Catalog No. NC2386557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.