Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodoadenosine 98.0+%, TCI America™
SDP

Catalog No. I07595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07595G 5 g
I0759200MG 200 mg
I07591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I07595G Supplier TCI America Supplier No. I07595G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35109-88-7
Molecular Formula C10H12IN5O4
Molecular Weight (g/mol) 393.141
MDL Number MFCD01320413
InChI Key MGEBVSZZNFOIRB-UUOKFMHZSA-N
Synonym 6-Amino-2-iodo-9-(beta-D-ribofuranosyl)purine
PubChem CID 169662
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-iodopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2N)I

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 5 g
Formula Weight 393.14
Percent Purity ≥98.0% (HPLC,N)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodoadenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.