Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobenzoic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620932 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-932 100 g
18-620-931 25 g
18-620-933 500 g
18-620-934 5 kg
18-620-935 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-932 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I2100100GM
Add to Cart
Add to Cart

Synonym: o-Iodobenzoic Acid, Iodobenzoic Acid, 2-Iodo Benzoic Acid

Chemical Identifiers

CAS 88-67-5
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.02
InChI Key CJNZAXGUTKBIHP-UHFFFAOYSA-N
IUPAC Name 2-iodobenzoic acid
SMILES OC(=O)C1=CC=CC=C1I

Specifications

Quantity 100 g
Percent Purity 98%
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.