Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. I0053100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0053100G 100 g
I005325G 25 g
I0053500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0053100G Supplier TCI America Supplier No. I0053100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-67-5
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD00002419
InChI Key CJNZAXGUTKBIHP-UHFFFAOYSA-N
Synonym o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure
PubChem CID 6941
ChEBI CHEBI:287979
IUPAC Name 2-iodobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I

Specifications

Melting Point 162°C
Quantity 100 g
Formula Weight 248.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodobenzoic Acid
EINECSNumber (3)-3578
RTECSNumber DH2975000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.