Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. I085825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I085825G 25 g
I08585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I085825G Supplier TCI America Supplier No. I085825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5159-41-1
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00004610
InChI Key WZCXOBMFBKSSFA-UHFFFAOYSA-N
Synonym 2-iodobenzyl alcohol, 2-iodophenyl methanol, o-iodobenzyl alcohol, 2-iodobenzylalcohol, benzenemethanol, 2-iodo, 2-iodobenzylic alcohol, 2-jod-benzylalkohol, o-iodo benzylalcohol, 2-iodo-benzyl alcohol
PubChem CID 107629
IUPAC Name (2-iodophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)I

Specifications

Melting Point 92°C
Color White
Boiling Point 145°C
Quantity 25 g
Formula Weight 234.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.