Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodobiphenyl 98.0+%, TCI America™
SDP

Catalog No. I096725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I096725G 25 g
I09675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I096725G Supplier TCI America Supplier No. I096725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2113-51-1
Molecular Formula C12H9I
Molecular Weight (g/mol) 280.108
MDL Number MFCD00039396
InChI Key QFUYDAGNUJWBSM-UHFFFAOYSA-N
Synonym 2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo
PubChem CID 75025
IUPAC Name 1-iodo-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2I

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 280.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodobiphenyl
RTECSNumber DV5384800
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.