Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iododibenzothiophene 98.0+%, TCI America™
SDP

Catalog No. I09731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09731G 1 g
I0973200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09731G Supplier TCI America Supplier No. I09731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 177586-41-3
Molecular Formula C12H7IS
Molecular Weight (g/mol) 310.15
MDL Number MFCD28975116
InChI Key LCTVJYHYWSCJAF-UHFFFAOYSA-N
PubChem CID 21709598
IUPAC Name 4-iodo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
SMILES IC1=CC2=C(SC3=C2C=CC=C3)C=C1

Specifications

Melting Point 88°C
Color White-Yellow
Quantity 1 g
Formula Weight 310.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iododibenzothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.