Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodoethanol (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. I039125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I039125G 25 g
I03915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I039125G Supplier TCI America Supplier No. I039125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
PubChem CID 12225
IUPAC Name 2-iodoethan-1-ol
SMILES OCCI

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
UN Number 2810
Formula Weight 171.97
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodoethanol (stabilized with Copper chip)
RTECSNumber KL3820000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.