Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodophenol, 98%

Catalog No. AAA1359914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1359914 25 g
AAA1359906 5 g
AAA1359922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1359914 Supplier Thermo Scientific Chemicals Supplier No. A1359914
Add to Cart

CAS: 533-58-4 | C6H5IO | 220.01 g/mol

2-Iodophenol is used in the synthesis of aryl 2-benzofuranyl and aryl 2-indolyl carbinols of high enantiomeric purity and 3,3-disubstituted-2,3-dihydrobenzofurans. Coupling with phenylacetylene in the presence of a Pd complex and CuI gives 2-phenylbenzofuran in good yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodophenol is used in the synthesis of aryl 2-benzofuranyl and aryl 2-indolyl carbinols of high enantiomeric purity and 3,3-disubstituted-2,3-dihydrobenzofurans. Coupling with phenylacetylene in the presence of a Pd complex and CuI gives 2-phenylbenzofuran in good yield.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-58-4
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.01
MDL Number MFCD00013963
InChI Key KQDJTBPASNJQFQ-UHFFFAOYSA-N
Synonym o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol
PubChem CID 10784
ChEBI CHEBI:16706
IUPAC Name 2-iodophenol
SMILES OC1=CC=CC=C1I

Specifications

Melting Point 39°C to 41°C
Density 1.947
Boiling Point 186°C to 187°C (160 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1855300
Sensitivity Light sensitive
Merck Index 14,5035
Solubility Information Soluble in water.
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 2-Iodophenol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 208-569-9
RTECSNumber SL5500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.