Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodothiophene 98.0+%, TCI America™
SDP

Catalog No. I0374100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0374100G 100 g
I037425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0374100G Supplier TCI America Supplier No. I0374100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3437-95-4
Molecular Formula C4H3IS
Molecular Weight (g/mol) 210.032
MDL Number MFCD00005424
InChI Key ROIMNSWDOJCBFR-UHFFFAOYSA-N
Synonym thiophene, 2-iodo, 2-thienyl iodide, 2-iodo-thiophene, .alpha.-iodothiophene, iodothiophene, thiophene, iodo, alpha-iodothiophene, a-iodothiophene, thiophene,iodo, 2-iodo thiophene
PubChem CID 18921
IUPAC Name 2-iodothiophene
SMILES C1=CSC(=C1)I

Specifications

Melting Point -40°C
Color Green-Yellow
Boiling Point 182°C
Quantity 100 g
Formula Weight 210.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.