Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodotoluene (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. I0217250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0217250G 250 g
I021725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0217250G Supplier TCI America Supplier No. I0217250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 615-37-2
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001042
InChI Key RINOYHWVBUKAQE-UHFFFAOYSA-N
Synonym 2-iodotoluene, o-iodotoluene, o-tolyl iodide, benzene, 1-iodo-2-methyl, o-methyliodobenzene, toluene, o-iodo, iodotoluene, ortho-iodotoluene, 2-methyliodobenzene, unii-8ok4h85t07
PubChem CID 5128
ChEBI CHEBI:39813
IUPAC Name 1-iodo-2-methylbenzene
SMILES CC1=CC=CC=C1I

Specifications

Boiling Point 211°C
Quantity 250 g
Formula Weight 218.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodotoluene (stabilized with Copper chip)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.