Learn More
Sigma Organic Chemistry 2-Isocyanatoethyl methacrylate | 5ML | 30674-80-7 | MFCD00080536 | 0.98
Supplier: Sigma Organic Chemistry 4770605ML

2-Isocyanatoethyl methacrylate, 5ML
About This Item:
Synonym(s): 2-(Methacryloyloxy)ethyl isocyanate; IEM
Linear Formula: H2C=C(CH3)CO2CH2CH2NCO
Molecular Weight: 155.15
CAS Number: 30674-80-7
MDL Number: MFCD00080536
UNSPSC Code: 12162002
Purity: 0.98
Boiling Point: 211 C (lit.)
Melting Point: -45 C (lit.)
Density: 1.098 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.45 (lit.)
Storage: 2-8C
EINECS Number: 250-284-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.