Learn More
Sigma Aldrich Fine Chemicals Biosciences Agarose Type I low EEO1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A6013
Encompass
Agarose is a polymer of agarobiose (L- and D-galactose) subunits. It is isolated from the seaweed Gelidium and Gracilaria. Agarose forms non-covalent network of bundles. It is exploited for the separation of nucleic acids. The concentration and pore size of the gel are inversely related. Agarose has wide range of separation and is simple and rapid to prepare without the need of catalyst.
Catalog No. 50-290-6697
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.