Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropenylnaphthalene 98.0+%, TCI America™
SDP

Catalog No. I041325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I041325G 25 g
I04135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I041325G Supplier TCI America Supplier No. I041325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3710-23-4
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00191524
InChI Key ANCUXNXTHQXICN-UHFFFAOYSA-N
PubChem CID 77301
IUPAC Name 2-(prop-1-en-2-yl)naphthalene
SMILES CC(=C)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 55°C
Color White-Yellow
Boiling Point 155°C
Quantity 25 g
Formula Weight 168.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Isopropenylnaphthalene
EINECSNumber (4)-0313
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.