Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropylanisole, 98%

Catalog No. p-7035664
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5338306 5 g
AAH5338314 25 g
AAH5338322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5338306 Supplier Thermo Scientific Chemicals Supplier No. H5338306
Only null left
Add to Cart

CAS: 2944-47-0 | C10H14O | 150.221 g/mol

2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropylanisole is the startind material in thesynthesis of (±)-shonanyl methyl ether and (±)-ferruginyl methyl ether.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2944-47-0
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00798590
InChI Key NNZRVXTXKISCGS-UHFFFAOYSA-N
Synonym 1-isopropyl-2-methoxybenzene, 2-isopropylanisole, o-isopropylanisole, anisole, o-isopropyl, benzene, 1-methoxy-2-1-methylethyl, 2-o-methoxyphenyl propane, 1-methoxy-2-propan-2-yl benzene, isopropylanisol, isopropyl anisole, acmc-20ansj
PubChem CID 270623
IUPAC Name 1-methoxy-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1OC

Specifications

Density 0.936
Boiling Point 198°C to 200°C
Flash Point 86°C (187°F)
Refractive Index 1.508
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 150.22
Percent Purity 98%
Chemical Name or Material 2-Isopropylanisole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.