Learn More
Sigma Organic Chemistry 2-Isopropylphenoxy acet | 1G | 25141-58-6 | MFCD00463165
Supplier: Sigma Organic Chemistry OTV0001341G

2-Isopropylphenoxy acet , 1G
About this item:
CAS #: 25141-58-6
MDL #: MFCD00463165
Chemical Formula: C11H14O3
Molecular Weight: 194.23
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.