Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-5-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. M229925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M229925G 25 g
M22995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M229925G Supplier TCI America Supplier No. M229925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76041-72-0
Molecular Formula C6H4F3NS
Molecular Weight (g/mol) 179.16
MDL Number MFCD00128893
InChI Key YFSCXLSUZZXNHW-UHFFFAOYSA-N
Synonym 2-mercapto-5-trifluoromethyl pyridine, 5-trifluoromethyl pyridine-2 1h-thione, 5-trifluoromethyl pyridine-2-thiol, 2-mercapto-5-trifluoromethylpyridine, 2 1h-pyridinethione, 5-trifluoromethyl, 5-trifluoromethyl-2-pyridinethiol, 5-trifluoromethyl-2 1h-pyridinethione, 5-trifluoromethyl-1h-pyridine-2-thione, 5-trifluoromethyl pyridin-2-thiol, 5-trifluoromethyl-2-mercaptopyridine
PubChem CID 2775246
IUPAC Name 5-(trifluoromethyl)-1H-pyridine-2-thione
SMILES C1=CC(=S)NC=C1C(F)(F)F

Specifications

Melting Point 150°C
Color Yellow
Quantity 25 g
Formula Weight 179.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-5-(trifluoromethyl)pyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.