Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptoethanol, 99%, pure

Catalog No. AC125470010
Change view
Click to view available options
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125470010 1 L Glass bottle
AC125470100 10 mL Glass bottle
AC125472500 250 mL Glass bottle
AC125470025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC125470010 Supplier Thermo Scientific Chemicals Supplier No. 125470010
Add to Cart

CAS: 60-24-2 | C2H6OS | 78.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
SMILES OCCS

Specifications

Melting Point -100.0°C
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 157.0°C to 158.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4990 to 1.5020
Linear Formula HSCH2CH2OH
Quantity 1 L
Beilstein 01,47
Fieser 01,643
Merck Index 15,5936
Specific Gravity 1.11
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,ether,benzene,soluble in acetone,miscible with most common organic solvents
IUPAC Name 2-sulfanylethan-1-ol
Viscosity 3.43 mPa.s (20°C)
Formula Weight 78.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause an allergic skin reaction.
May cause damage to organs through prolon
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 200-464-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.