Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Mercaptoethyl)pyrazine 98.0+%, TCI America™
SDP

Catalog No. M18451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18451G 1 g
M18455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18451G Supplier TCI America Supplier No. M18451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35250-53-4
Molecular Formula C6H8N2S
Molecular Weight (g/mol) 140.204
MDL Number MFCD00053160
InChI Key QKVWBAMZPUHCMO-UHFFFAOYSA-N
Synonym pyrazineethanethiol, 2-pyrazinylethanethiol, 2-pyrazineethanethiol, pyrazine ethanethiol, pyrazinyl ethanethiol, 2-2-mercaptoethyl pyrazine, 2-pyrazinylethylmercaptan, 2-mercaptoethyl pyrazine, mercaptoethylpyrazine, unii-f7i6g0e56f
PubChem CID 61945
IUPAC Name 2-pyrazin-2-ylethanethiol
SMILES C1=CN=C(C=N1)CCS

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
UN Number 2810
Formula Weight 140.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Mercaptoethyl)pyrazine
RTECSNumber KJ2551000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.