Learn More
Bis(2-methoxyethyl) ether, 99%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -64.0°C |
| Density | 0.9370g/mL |
| Boiling Point | 162.0°C |
| Flash Point | 51°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Plastic drum |
| Refractive Index | 1.4070 to 1.4090 |
| Linear Formula | (CH3OCH2CH2)2O |
| Quantity | 10 L |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May damage fertility or the unborn child. Flammable liquid and vapor. May form explosive peroxides. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. |
| EINECSNumber | 203-924-4 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.