Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopropionic acid, 95%

Catalog No. AC138790025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138790025 2.5 kg Glass bottle
AC138791000 100 g Glass bottle
AC138795000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138790025 Supplier Thermo Scientific Chemicals Supplier No. 138790025
Only null left
Add to Cart

CAS: 79-42-5 | C3H6O2S | 106.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-42-5
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004862
InChI Key PMNLUUOXGOOLSP-UHFFFAOYNA-N
Synonym 2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
PubChem CID 62326
ChEBI CHEBI:47872
IUPAC Name 2-sulfanylpropanoic acid
SMILES CC(S)C(O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 10°C to 14°C
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 102°C (16.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass bottle
Refractive Index 1.4804 to 1.4824
Linear Formula CH3CH(SH)CO2H
Quantity 2.5 kg
Beilstein 03, 289
Fieser 15,307
Specific Gravity 1.19
Merck Index 15, 9493
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with alcohol,ether and acetone,miscible with benzol,petroleumether,chloroform,,chloroethane,diethylether
Formula Weight 106.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Mercaptopropionic acid, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 201-206-5
RTECSNumber UF5250000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.