Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercaptopyridine N-oxide sodium salt hydrate, 98%, Thermo Scientific™

Catalog No. p-7082052
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1230306 5 g
AAA1230314 25 g
AAA1230322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1230306 Supplier Thermo Scientific Chemicals Supplier No. A1230306

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304675-78-3
Molecular Formula C5H4NNaOS
Molecular Weight (g/mol) 149.14
MDL Number MFCD00151244
InChI Key WNGMMIYXPIAYOB-UHFFFAOYSA-M
Synonym sodium 2-mercaptopyridine n-oxide hydrate
PubChem CID 43835086
IUPAC Name sodium (1-oxidopyridin-1-ium-2-yl)sulfanide
SMILES [Na+].[O-][N+]1=CC=CC=C1[S-]

Specifications

Quantity 5 g
Merck Index 14,7994
Formula Weight 149.15 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyridine N-oxide sodium salt hydrate
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber UT9000000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.