Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mesitylenesulfonyl Hydrazide 97.0+%, TCI America™
SDP

Catalog No. M119825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M119825G 25 g
M11985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M119825G Supplier TCI America Supplier No. M119825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16182-15-3
Molecular Formula C9H14N2O2S
Molecular Weight (g/mol) 214.283
MDL Number MFCD00007585
InChI Key JQUBKTQDNVZHIY-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide
PubChem CID 85321
IUPAC Name 2,4,6-trimethylbenzenesulfonohydrazide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C

Specifications

Melting Point 114°C
Color White
Quantity 25 g
UN Number 1325
Formula Weight 214.28
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mesitylenesulfonyl Hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.