Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-2-(1-naphthyl)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. M13685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13685G Supplier TCI America Supplier No. M13685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63628-25-1
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
MDL Number MFCD03093635
InChI Key YVWMPILNFZOQSZ-UHFFFAOYSA-N
Synonym alpha-Methoxy-alpha-methyl-1-naphthaleneacetic Acid
PubChem CID 11218470
IUPAC Name 2-methoxy-2-naphthalen-1-ylpropanoic acid
SMILES CC(C1=CC=CC2=CC=CC=C21)(C(=O)O)OC

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 5 g
Formula Weight 230.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-2-(1-naphthyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.