Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M231125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2311-25G 25 g
M23115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2311-25G Supplier TCI America Supplier No. M231125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 127972-00-3
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.98
MDL Number MFCD02179465
InChI Key CSVKZOZMPSRLTC-UHFFFAOYSA-N
PubChem CID 2773485
IUPAC Name (2-methoxy-5-methylphenyl)boronic acid
SMILES COC1=CC=C(C)C=C1B(O)O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 25 g
Synonym 2-methoxy-5-methylphenyl boronic acid, 2-methoxy-5-methylbenzeneboronic acid, 2-methoxy-5-methyl-phenyl boronic acid, 2-methoxy-5-methylphenyl boranediol, 5-methyl-2-methoxyphenylboronic acid
Formula Weight 165.98
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.