Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-pyrimidylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193761 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23351G 1 g
M2335-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23351G Supplier TCI America Supplier No. M23351G
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 2-Methoxy-5-pyrimidylboronic Acid (contains varying amounts of Anhydride)
CAS 628692-15-9
Melting Point 161°C
Color Yellow
Molecular Formula C5H7BN2O3
Quantity 1 g
MDL Number MFCD03094664
Synonym 2-methoxypyrimidine-5-boronic acid, 2-methoxypyrimidin-5-yl boronic acid, 2-methoxy-5-pyrimidineboronic acid, 2-methoxypyrimidine-5-boronicacid, 2-methoxy-pyrimidine-5-boronic acid, 2-methoxypyrimidin-5-yl-5-boronic acid, 2-methoxyl-5-pyrimidylboric acid, 2-methoxy-5-pyrimidylboronic acid, 2-methoxypyrimidin-5-yl boranediol, 2-methoxy-5-pyrimidinylboronic acid
InChI Key YPWAJLGHACDYQS-UHFFFAOYSA-N
SMILES B(C1=CN=C(N=C1)OC)(O)O
IUPAC Name (2-methoxypyrimidin-5-yl)boronic acid
Molecular Weight (g/mol) 153.932
PubChem CID 2736778
Formula Weight 153.93
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.