Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-(trifluoromethyl)aniline 98.0+%, TCI America™
SDP

Catalog No. M221225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M221225G 25 g
M22125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M221225G Supplier TCI America Supplier No. M221225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 349-65-5
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00042486
InChI Key RKUSRLUGUVDNKP-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethyl aniline, 3-amino-4-methoxybenzotrifluoride, 2-amino-4-trifluoromethylanisole, 2-methoxy-5-trifluoromethyl-aniline, 2-methoxy-5-trifluoromethyl-phenylamine, 2-methoxy-5-trifluoromethyl phenylamine, benzenamine, 2-methoxy-5-trifluoromethyl, pubchem2720, acmc-209ic5, 4-trifluromethyl-o-anisidine
PubChem CID 600637
IUPAC Name 2-methoxy-5-(trifluoromethyl)aniline
SMILES COC1=C(C=C(C=C1)C(F)(F)F)N

Specifications

Melting Point 60°C
Quantity 25 g
Formula Weight 191.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.