Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-6-methylaniline 98.0+%, TCI America™
SDP

Catalog No. M180425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M180425G 25 g
M18045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M180425G Supplier TCI America Supplier No. M180425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50868-73-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00075455
InChI Key HKOJYPPTIPJZAZ-UHFFFAOYSA-N
Synonym 6-Methoxy-o-toluidine, 2-Amino-1-methoxy-3-methylbenzene
PubChem CID 459249
IUPAC Name 2-methoxy-6-methylaniline
SMILES COC1=CC=CC(C)=C1N

Specifications

Color Brown-White
Boiling Point 116°C
Quantity 25 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methoxy-6-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.