Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-6-(tri-n-butylstannyl)pyrazine, 95%

Catalog No. p-7034008
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51528MD 250 mg
AAH5152803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51528MD Supplier Thermo Scientific Chemicals Supplier No. H51528MD
Only null left
Add to Cart

CAS: 1105511-66-7 | C17H32N2OSn | 399.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1105511-66-7
Molecular Formula C17H32N2OSn
Molecular Weight (g/mol) 399.166
MDL Number MFCD08275735
InChI Key WWRXJELRWGUPKZ-UHFFFAOYSA-N
Synonym 2-methoxy-6-tributylstannyl pyrazine, 2-tri-n-butylstannyl-6-methoxypyrazine, 2-methoxy-6-tributylstannanyl pyrazine, 2-methoxy-6-tri-n-butylstannyl pyrazine, 2-tributylstannyl-6-methoxypyrazine
PubChem CID 16427103
IUPAC Name tributyl-(6-methoxypyrazin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC(=CN=C1)OC

Specifications

Quantity 250 mg
UN Number UN2788
Formula Weight 399.15
Percent Purity 95%
Chemical Name or Material 2-Methoxy-6-(tri-n-butylstannyl)pyrazine
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.