Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-beta-estradiol 97.0+%, TCI America™
SDP

Catalog No. M2530100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2530100MG 100 mg
M253025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2530100MG Supplier TCI America Supplier No. M2530100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 362-07-2
Molecular Formula C19H26O3
Molecular Weight (g/mol) 302.414
MDL Number MFCD00010489
InChI Key CQOQDQWUFQDJMK-SSTWWWIQSA-N
Synonym 2-methoxyestradiol, panzem, 2-methoxyestradiol-17beta, 2-hydroxyestradol 2-methyl ether, estradiol, 2-methoxy, 2-hydroxyestradiol 2-methyl ether, pulmolar, 2-methoxyestra-1,3,5 10-triene-3,17beta-diol, panzem ncd, 2-methoxy-17beta-estradiol
PubChem CID 66414
ChEBI CHEBI:28955
IUPAC Name (8R,9S,13S,14S,17S)-2-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=CC(=C(C=C34)OC)O

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 100 mg
Formula Weight 302.41
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-beta-estradiol
RTECSNumber KG7537500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.