Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Methoxyacetophenone 97.0+%, TCI America™
SDP

Catalog No. M1445250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1445250G 250 g
M144525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1445250G Supplier TCI America Supplier No. M1445250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 579-74-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008725
InChI Key DWPLEOPKBWNPQV-UHFFFAOYSA-N
Synonym 2'-methoxyacetophenone, 1-2-methoxyphenyl ethanone, o-methoxyacetophenone, ethanone, 1-2-methoxyphenyl, o-acetylanisole, 1-2-methoxyphenyl ethan-1-one, o-acetanisole, 2-acetylanisole, 2-methyoxyacetophenone oxime, ortho-methoxyacetophenone
PubChem CID 68481
IUPAC Name 1-(2-methoxyphenyl)ethan-1-one
SMILES COC1=CC=CC=C1C(C)=O

Specifications

Color Yellow
Quantity 250 g
Formula Weight 150.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Methoxyacetophenone
EINECSNumber (3)-2676
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.