Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybenzeneboronic acid, 97%

Catalog No. AAB2107106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2107106 5 g
AAB2107103 1 g
AAB2107114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2107106 Supplier Thermo Scientific Chemicals Supplier No. B2107106
Add to Cart

CAS: 5720-06-9 | C7H9BO3 | 151.96 g/mol

2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5720-06-9
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00236047
InChI Key ROEQGIFOWRQYHD-UHFFFAOYSA-N
Synonym 2-methoxybenzeneboronic acid, 2-methoxyphenyl boronic acid, o-methoxyphenylboronic acid, 2-methoxyphenyl boranediol, 2-boronoanisole, 2-methoxypenylboronic acid, o-methoxyphenyl boronic acid, 2-methoxy phenylboronic acid, 2-methoxybenzene boronic acid
PubChem CID 2733958
IUPAC Name (2-methoxyphenyl)boronic acid
SMILES COC1=CC=CC=C1B(O)O

Specifications

Melting Point 102°C to 106°C
Quantity 5 g
Beilstein 3030981
Solubility Information Insoluble in water.
Formula Weight 151.96
Percent Purity 97%
Chemical Name or Material 2-Methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.