Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxybenzenethiol 98.0+%, TCI America™
SDP

Catalog No. M167025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M167025G 25 g
M16705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M167025G Supplier TCI America Supplier No. M167025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7217-59-6
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
InChI Key DSCJETUEDFKYGN-UHFFFAOYSA-N
Synonym 2-methoxythiophenol, thioguaiacol, benzenethiol, 2-methoxy, o-methoxythiophenol, o-methoxybenzenethiol, 2-methoxy thiophenol, 2-mercaptoanisole, 2-methoxybenzene-1-thiol, 2-methoxy-benzenethiol, benzenethiol, o-methoxy
PubChem CID 23642
IUPAC Name 2-methoxybenzenethiol
SMILES COC1=CC=CC=C1S

Specifications

Color Yellow
Boiling Point 219°C
Quantity 25 g
Formula Weight 140.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzenethiol
RTECSNumber DC1790000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.