Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methoxycarbonyl)-6-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. M279525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M279525G 25 g
M27955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M279525G Supplier TCI America Supplier No. M279525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21606-04-2
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.156
MDL Number MFCD00034781
InChI Key DJMQLZPEBHSABD-UHFFFAOYSA-N
PubChem CID 4399477
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 25 g
Formula Weight 225.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methoxycarbonyl)-6-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.