Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol, ACS, 99.3+%

Catalog No. AA31733K7
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA31733K7 4 L
AA31733M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AA31733K7 Supplier Thermo Scientific Chemicals Supplier No. 031733K7
Add to Cart
This item is not returnable. View return policy

CAS: 109-86-4 | C3H8O2 | 76.095 g/mol

2-Methoxy ethanol is used as solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Melting Point -85°C
Color Colorless
Density 0.965
pH 4 to 7
Boiling Point 124°C to 125°C
Flash Point 46°C (115°F)
Odor Characteristic
Refractive Index 1.4025
Linear Formula CH3OCH2CH2OH
Quantity 4 L
UN Number UN1188
Beilstein 1731074
Merck Index 14,6038
Solubility Information Miscible with water,alcohol,glycerol,ether,acetone,DMFMiscible with water,alcohol,ether,acetone,hydrocarbons,ketones,glycols and dimethylformamide.
Formula Weight 76.1
Percent Purity ≥99.3%
Grade ACS
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
Hazard Category H226-H302+H312+H332-H360FD-H370-H373
Hazard Statement GHS H Statement
H360-H226-H302-H312-H332;
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P307+P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
EINECSNumber 203-713-7
RTECSNumber KL5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.