Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol, ACS reagent

Catalog No. AC447480010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447480010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC447480010 Supplier Thermo Scientific Chemicals Supplier No. 447480010
  • $92.00
    $73.65 / Each of 1

    Save $18.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-86-4 | C3H8O2 | 76.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.09
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Melting Point -85.0°C
Density 0.9600g/mL
Boiling Point 124.0°C
Flash Point 38°C
Refractive Index 1.402
Linear Formula CH3OCH2CH2OH
Quantity 1 L
Merck Index 15,6063
Specific Gravity 0.96
Solubility Information Solubility in water: soluble
Viscosity 1.7 mPa.s (20°C)
Formula Weight 76.09
Grade ACS Reagent
Titratable Acid 0.002 meq/g max
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May damage fertility.
May damage the unborn child.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/sh
EINECSNumber 203-713-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.